Lithium dichromate hydrate, 98+% 1 * 250 g
Catalog # 47394.30
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
Diethylene glycol monomethyl ether, Sigma-Aldrich®
Supplier: Merck
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
Supplier: PanReac AppliChem
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Ethanol denatured absolute ≥98% (by GC) (denaturated with 1% MEK and 0.001% Bitrex®)
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised ACS
Supplier: Thermo Fisher Scientific
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific