SARS-COV-2 SPIKE REAGENT 1 * 100 AU
Catalog # CAYM502221-100
Supplier: Cayman Chemical
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol (untaxed), anhydrous, tested according to Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals