SARAFLOXACIN-D8 (HYDROCHLORIDE) 1 * 5 mg
CAYM34226-5
Frequently Bought Together
Butyl acetate ≥99.5% for HPLC
: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
2-Propanol 99.5% for HPLC
: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
: Thermo Fisher Scientific
Cyclohexanone, Sigma-Aldrich®
: Merck
tert-Butanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
2-Methylpentane ≥99%
: Thermo Fisher Scientific
2-Ethoxyethanol 99%, Reagent Grade
: Honeywell Chemicals