4-Nitrophenyl Phosphate (sodium salt hydrate) 1 * 50 g
Catalog # CAYM34081-50
Supplier: Cayman Chemical
4-Nitrophenyl Phosphate (sodium salt hydrate) 1 * 50 g
Catalog # CAYM34081-50
Supplier: Cayman Chemical
Frequently Bought Together
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Water, extra pure, Deionized
Supplier: Thermo Fisher Scientific
Acetone, for GC Plus, Burdick & Jackson™
Supplier: Honeywell Chemicals
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
(±)-2-Ethyl-1-hexanol 99%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific