4-Nitrophenyl Phosphate (sodium salt hydrate) 1 * 5 g
Catalog # CAYM34081-5
Supplier: Cayman Chemical
4-Nitrophenyl Phosphate (sodium salt hydrate) 1 * 5 g
Catalog # CAYM34081-5
Supplier: Cayman Chemical
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
n-Octane ≥98%
Supplier: Thermo Fisher Scientific