Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
5-Bromo-4-chloro-3-indolyl--D-Glucuronide (cyclohexylammonium salt hydrate) 1 * 25 mg
Catalog # CAYM33591-25
Supplier:
Cayman Chemical
Expand image
5-Bromo-4-chloro-3-indolyl--D-Glucuronide (cyclohexylammonium salt hydrate) 1 * 25 mg
Catalog # CAYM33591-25
Supplier:
Cayman Chemical
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals
Methanol ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
n-Pentane ≥95%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol 70 % pure 1 * 1 L
Supplier: PanReac AppliChem
Peptide and protein MALDI-MS calibration kit, ProteoMass™, Supelco®
Supplier: Merck