3,3′-Dithiodipropionic Acid di(N-hydroxysuccinimide ester) 1 * 500 mg
Katalogové číslo CAYM33405-500
Dodavatel: Cayman Chemical
3,3′-Dithiodipropionic Acid di(N-hydroxysuccinimide ester) 1 * 500 mg
Katalogové číslo CAYM33405-500
Dodavatel: Cayman Chemical
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
Dodavatel: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Dodavatel: EPPENDORF
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Dodavatel: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM®, super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Transfer pipettes
Dodavatel: RATIOLAB
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Dodavatel: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Dodavatel: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Dodavatel: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Dodavatel: VWR Collection
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Dodavatel: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals