3,3′-Dithiodipropionic Acid di(N-hydroxysuccinimide ester) 1 * 250 mg
CAYM33405-250
3,3′-Dithiodipropionic Acid di(N-hydroxysuccinimide ester) 1 * 250 mg
CAYM33405-250
Frequently Bought Together
Chlorobenzene ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Isoamyl alcohol, TECHNICAL
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals