RU-24969 (SUCCINATE) 1 * 50 mg
Catalog # CAYM33142-50
Supplier: Cayman Chemical
Frequently Bought Together
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
[E]120ML CLICKSEAL CONTAINER W/30ML FILL 1 * 1 MJ
Supplier: ACS KITTING
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals