(4-Carboxybutyl-d4)triphenylphosphonium (bromide) 1 * 25 mg
Catalog # CAYM31254-25
Supplier: Cayman Chemical
(4-Carboxybutyl-d4)triphenylphosphonium (bromide) 1 * 25 mg
Catalog # CAYM31254-25
Supplier: Cayman Chemical
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Supplier: VWR Collection
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals