HYDROXY-PEG2 ACID 1 * 250 mg
Katalogové číslo CAYM30903-250
Dodavatel: Cayman Chemical
Frequently Bought Together
2-Propanol ≥99.0% USP, Ph. Eur.
Dodavatel: VWR Chemicals
Screw caps for laboratory/media bottles
Dodavatel: SciLabware
Serological pipettes, Stripette™
Dodavatel: Corning
N,N-Dimethylhydrazine
Dodavatel: Merck
Glycerine, Sigma-Aldrich®
Dodavatel: Merck
n-Hexane
Dodavatel: Lach-ner S.R.O.
Acetone ≥99.5%, Environmental Grade
Dodavatel: Thermo Fisher Scientific
Pyridine, extra dry 99.5%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
Advanced TC™ cell culture flasks
Dodavatel: Greiner Bio-One
11 components: 100mg/l each of Li Mo Ni Sb Be Co Pb V Sn Mg Ti in HNO3 5% 1 * 100 mL
Dodavatel: CUSTOM MADE CHEMICALS LAB
Ice buckets with lids, Azlon®
Dodavatel: SciLabware
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with screw caps, Axygen®
Dodavatel: Corning
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Dodavatel: Thermo Fisher Scientific