3-HYDROXY OCTANOIC ACID 1 * 5 mg
Catalog # CAYM24609-5
Supplier: Cayman Chemical
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Serological pipettes, Stripette™
Supplier: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals