9-HYDROXY STEARIC ACID 1 * 25 mg
Katalogové číslo CAYM18016-25
Dodavatel: Cayman Chemical
Frequently Bought Together
PCR microtubes
Dodavatel: EPPENDORF
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Dodavatel: BIOCISION
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
Dodavatel: VWR Chemicals
VWR®, Cryoboxes, Cardboard, 133×133 mm
Dodavatel: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Dodavatel: VWR Collection
Screw cap microcentrifuge tubes, SuperClear®
Dodavatel: LABCON
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Dodavatel: Avantor Fluid Handling
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Dodavatel: Thermo Fisher Scientific
VWR®, Cryo Boxes
Dodavatel: VWR Collection
Cryoboxes, 133×133 mm
Dodavatel: TENAK
Dichloromethane ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Cryogenic tubes
Dodavatel: Brand
Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Dodavatel: Honeywell Chemicals