Titanium sub-hydride (TiH0.65), -325 mesh, 99%, packed under argon 1 * 25 g
Katalogové číslo 47320.14
Supplier: Thermo Fisher Scientific
Titanium sub-hydride (TiH0.65), -325 mesh, 99%, packed under argon 1 * 25 g
Katalogové číslo 47320.14
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Cyclohexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
Supplier: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Toluene, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific