Titanium sub-hydride (TiH1.65), -325 mesh, 99%, packed under argon 1 * 500 g
Catalog # 47302.36
Supplier: Thermo Fisher Scientific
Titanium sub-hydride (TiH1.65), -325 mesh, 99%, packed under argon 1 * 500 g
Catalog # 47302.36
Supplier: Thermo Fisher Scientific
Frequently Bought Together
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific