Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Yeast Extract Agar

Yeast Extract Agar

Yeast extract agar, 600 ml, Bottle, 600 mL
Katalogové číslo LIOF403120
Dodavatel:   LIOFILCHEM
Yeast Extract Agar
Katalogové číslo LIOF403120
Dodavatel:  LIOFILCHEM
Číslo CAS:  
Yeast Extract Agar

Yeast extract agar, 600 ml, Bottle, 600 mL

Přejít na:
Číslo CAS:  
Price
1 230,00 Kč
Přihlásit se pro dostupnost
1 * 600 mL
Product Details & Documents

Medium for microorganisms enumeration in water.

Certifications: ISO6222 certified.

Specifikace

Specifikace

  • Pack type:
    Bottle
  • Pk:
    600 mL
  • Description:
    Yeast extract agar
  • Volume:
    600 ml

Specifikace

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.