ContactUsLinkComponent
=99.9%, acid <10 ppm, H2O <10 ppm CAS Number: 616-38-6 Formula: C3H6O3 Formula Weight: 90.08 g/mol Qualitätsniveau 100 Dampfdichte 3.1 (vs air) Dampfdruck 18 mmHg ( 21.1 °C) Assay =99.9% Form liquid Brechungsindex n20/D 1.368 (lit.) bp 90 °C (lit.) m 1 * 500 g
Catalog # SIAL809942-500G
Supplier:  Merck
=99.9%, acid <10 ppm, H2O <10 ppm CAS Number: 616-38-6 Formula: C3H6O3 Formula Weight: 90.08 g/mol Qualitätsniveau 100 Dampfdichte 3.1 (vs air) Dampfdruck 18 mmHg ( 21.1 °C) Assay =99.9% Form liquid Brechungsindex n20/D 1.368 (lit.) bp 90 °C (lit.) m 1 * 500 g
Catalog # SIAL809942-500G
Supplier:  Merck
Frequently Bought Together