A single, non-glycosylated biologically active polypeptide chain corresponding to 132 residues of TSLP. 1 * 100 µG
Catalog # NOVUNBP2-35083-100
Supplier: Novus Biologicals
A single, non-glycosylated biologically active polypeptide chain corresponding to 132 residues of TSLP. 1 * 100 µG
Catalog # NOVUNBP2-35083-100
Supplier: Novus Biologicals
Frequently Bought Together
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Plugs for test tubes
Supplier: BOETTGER PAUL
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
Storage bottles, octogonal, PET, Corning® Gosselin™
Supplier: Corning
Petri dishes
Supplier: Greiner Bio-One
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Microplates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Sampling scoops, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Whatman™ Cellulose Blotting Papers, Grade GB003, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
NEOBOX 81 PLUS, UNTERTEIL BLAU 1 * 1 ST
Supplier: NEOLAB
VWR® Cryoboxes and Dividers
Supplier: VWR Collection
CapInsert™ cap inserts for Cryovial® tubes
Supplier: Simport Scientific