ContactUsLinkComponent
EvaRuby is the first visible red fluorogenic DNA dye for qPCR and HRM, with Ex/Em at 480/610 nm upon binding to dsDNA. It can be combined with fluorescent probes for probe-based qPCR with high resolution melt analysis in the same reaction. 20X in wat 1 * 500 µl
Catalog # 31079-T
Supplier:  Biotium
EvaRuby is the first visible red fluorogenic DNA dye for qPCR and HRM, with Ex/Em at 480/610 nm upon binding to dsDNA. It can be combined with fluorescent probes for probe-based qPCR with high resolution melt analysis in the same reaction. 20X in wat 1 * 500 µl
Catalog # 31079-T
Supplier:  Biotium
Frequently Bought Together