ContactUsLinkComponent
Pan monocytes were negatively selected via immunomagnetic separation from a leukopheresis collection. After isolation, the monocytes were tested for viability by NC-200 and purity by flow cytometry for CD14/CD16 expression. The monocytes were cryopr 1 * 25 Mou
Catalog # PB14/16NC-2
Pan monocytes were negatively selected via immunomagnetic separation from a leukopheresis collection. After isolation, the monocytes were tested for viability by NC-200 and purity by flow cytometry for CD14/CD16 expression. The monocytes were cryopr 1 * 25 Mou
Catalog # PB14/16NC-2
Frequently Bought Together