ContactUsLinkComponent
Hooded coverall combining Tyvek with a light SMS back panel from shoulders to ankle. Elasticated wrists, waist and ankles. Zipper flap. Thumbhole on sleeve. Bulk packed, no individual garment packaging reducing the waste. White. Antistatic. Size M 1 * 25 items
Catalog # DUPOTD0127MWH00
Supplier:  DuPont
undefined
Hooded coverall combining Tyvek with a light SMS back panel from shoulders to ankle. Elasticated wrists, waist and ankles. Zipper flap. Thumbhole on sleeve. Bulk packed, no individual garment packaging reducing the waste. White. Antistatic. Size M 1 * 25 items
Catalog # DUPOTD0127MWH00
Supplier:  DuPont