ContactUsLinkComponent
VISOCOLOR Powder Pillows Nitrite reagent set for photometric determination measuring range: 0.01-0.30 mg/L NO2-N 100 determinations __UN 3316 Chemical kit 9 II 0.015 KG ADR/GGVSEB M11 (E) ADR 3.3.1/251 PAX 960 10 KG CAO 960 10 KG Q: 0.002 / 0.002 1 * 1 KIT
Catalog # MANA936230
Supplier:  MACHEREY-NAGEL
VISOCOLOR Powder Pillows Nitrite reagent set for photometric determination measuring range: 0.01-0.30 mg/L NO2-N 100 determinations __UN 3316 Chemical kit 9 II 0.015 KG ADR/GGVSEB M11 (E) ADR 3.3.1/251 PAX 960 10 KG CAO 960 10 KG Q: 0.002 / 0.002 1 * 1 KIT
Catalog # MANA936230
Supplier:  MACHEREY-NAGEL
Frequently Bought Together