ContactUsLinkComponent
CHROMABOND QuEChERS Mix II acetate extraction mix (acc. to AOAC) consisting of: - 6000 mg MgSO4 - 1500 mg Na acetate individually we ighed and filled into 50 mL centrifuge tubes (PP) with screw cap (PE) pack of 50 1 * 50 mL
Catalog # MANA730971.1
Supplier:  MACHEREY-NAGEL
CHROMABOND QuEChERS Mix II acetate extraction mix (acc. to AOAC) consisting of: - 6000 mg MgSO4 - 1500 mg Na acetate individually we ighed and filled into 50 mL centrifuge tubes (PP) with screw cap (PE) pack of 50 1 * 50 mL
Catalog # MANA730971.1
Supplier:  MACHEREY-NAGEL
Frequently Bought Together