ContactUsLinkComponent
Vial & Cap Kit. 100 EA 1.8ml, Screw Top, Hydrophobic, Amber Autosampler Vials and matching Black Caps with Clear Silicone Rubber / P TFE Pre-Slit Septa. RSA-Pro X Brand. 1 PK. 1 * 100 items
Catalog # MICS9512S-3DAP-PX
Supplier:  MICROSOLV
Vial & Cap Kit. 100 EA 1.8ml, Screw Top, Hydrophobic, Amber Autosampler Vials and matching Black Caps with Clear Silicone Rubber / P TFE Pre-Slit Septa. RSA-Pro X Brand. 1 PK. 1 * 100 items
Catalog # MICS9512S-3DAP-PX
Supplier:  MICROSOLV
Frequently Bought Together