14 components 10mg/kg each of Ge-Nb -Pd-Pt-Rh-Sb-Sn-Ta-Ti-W-Zr-Ir-Os-Ru in HNO3 5% HCl 0.5% - HF 0.5% CRM, ISO 17034 and ISO 17 025 1 * 100 mL
Catalog # ACSDRM057049L1.2
Supplier: CPACHEM
14 components 10mg/kg each of Ge-Nb -Pd-Pt-Rh-Sb-Sn-Ta-Ti-W-Zr-Ir-Os-Ru in HNO3 5% HCl 0.5% - HF 0.5% CRM, ISO 17034 and ISO 17 025 1 * 100 mL
Catalog # ACSDRM057049L1.2
Supplier: CPACHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Nitrile Gloves
Supplier: VWR Collection
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
ICP METAL STANDARD 31 COMPONENT 1 * 100 mL
Supplier: CPACHEM