3-HYDROXY MYRISTIC ACID METHYL ESTER 1 * 25 mg
Catalog # CAYM24645-25
Supplier: Cayman Chemical
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Cleanroom overalls, Kimtech™ A5
Supplier: KIMBERLY CLARK
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
3-HYDROXY DECANOIC ACID METHYL ESTER 1 * 10 mg
Supplier: Cayman Chemical
Erlenmeyer flasks, baffled, disposable, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific