ContactUsLinkComponent
Lightning-Link antibody labeling kits enable the direct labeling of antibodies, proteins, peptides or other biomolecules for use in R &D applications, drug discovery and the development of diagnostic kits 1 * 1 mg
Catalog # NOVU705-0015
Supplier:  Novus Biologicals
Lightning-Link antibody labeling kits enable the direct labeling of antibodies, proteins, peptides or other biomolecules for use in R &D applications, drug discovery and the development of diagnostic kits 1 * 1 mg
Catalog # NOVU705-0015
Supplier:  Novus Biologicals
Frequently Bought Together