ContactUsLinkComponent
Sterile sampling bag - w x h: 11 x 25 cm / 4.3 x 10 pouces - Thickness: 70 µm / 3 mil - Capacity: 600 mL / 20 oz - Number of stickers: 1 1 * 500 items
Catalog # INSC145115
Supplier:  INTERSCIENCE
undefined
Sterile sampling bag - w x h: 11 x 25 cm / 4.3 x 10 pouces - Thickness: 70 µm / 3 mil - Capacity: 600 mL / 20 oz - Number of stickers: 1 1 * 500 items
Catalog # INSC145115
Supplier:  INTERSCIENCE