ContactUsLinkComponent
Sterile sampling bag - w x h: 11 x 20 cm / 4.3 x 8 pouces - Thickness: 70 µm / 3 mil - Capacity: 350 mL / 12 oz - Number of stickers: 1 1 * 500 items
Catalog # INSC145112
Supplier:  INTERSCIENCE
undefined
Sterile sampling bag - w x h: 11 x 20 cm / 4.3 x 8 pouces - Thickness: 70 µm / 3 mil - Capacity: 350 mL / 12 oz - Number of stickers: 1 1 * 500 items
Catalog # INSC145112
Supplier:  INTERSCIENCE