ContactUsLinkComponent
HelixMark® Standard Silicone Tubing Material: DuPont Liveo Q7-4750 OD: 7,95 mm, ID: 4,78 mm Packaging: 15 m in double plastic bag 1 * 15 m
Catalog # HELI45-630-122
HelixMark® Standard Silicone Tubing Material: DuPont Liveo Q7-4750 OD: 7,95 mm, ID: 4,78 mm Packaging: 15 m in double plastic bag 1 * 15 m
Catalog # HELI45-630-122
Frequently Bought Together