ContactUsLinkComponent
DENSITY STANDARD 1.2849 G/ML 20 ºC 1 * 60 mL
Catalog # PARGDEN20-13
Supplier:  PARAGON SCIENTIFIC
DENSITY STANDARD 1.2849 G/ML 20 ºC 1 * 60 mL
Catalog # PARGDEN20-13
Supplier:  PARAGON SCIENTIFIC