ContactUsLinkComponent
Glass filtration set VF3, complete: glass funnel 300ml, 47mm, glass suction cap with ST socket (support base), clamp, glass receiver flask 1000ml and fixing sucker 1 * 1 items
Catalog # WITG00829.16720003
Supplier:  witeg Labortechnik
Glass filtration set VF3, complete: glass funnel 300ml, 47mm, glass suction cap with ST socket (support base), clamp, glass receiver flask 1000ml and fixing sucker 1 * 1 items
Catalog # WITG00829.16720003
Supplier:  witeg Labortechnik
Frequently Bought Together