ContactUsLinkComponent
Tube, Specimen Collection and Transport Tube Labcon SuperClear® 10 mL Sterile Cap with integral molded elastomeric seal Includes molded graduations and writing surface ATP, RNase/DNase and Endotoxin Free 1 * 500 items
Catalog # LBCN3814-875-008-9
Supplier:  Labcon
Tube, Specimen Collection and Transport Tube Labcon SuperClear® 10 mL Sterile Cap with integral molded elastomeric seal Includes molded graduations and writing surface ATP, RNase/DNase and Endotoxin Free 1 * 500 items
Catalog # LBCN3814-875-008-9
Supplier:  Labcon
Frequently Bought Together