Gb/T 24153-2009 Nitrosamines Mixture 137 100 µg/Ml In Methanol 1 * 1 mL
Catalog # EHERA50000137ME
Supplier: EHRENSTORFER
Gb/T 24153-2009 Nitrosamines Mixture 137 100 µg/Ml In Methanol 1 * 1 mL
Catalog # EHERA50000137ME
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Capillary column ferrules for gas chromatography
Supplier: Agilent