Ucn2 ELISA Kit(Rat) is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for Ucn2 has been pre-coated onto a 96-wellplate (12 x 8 Well Strips) 1 * 1 KIT
Catalog # AVIVOKCD02959
Supplier: Aviva Systems Biology
Frequently Bought Together