0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Serological Pipette Graduated 5ml Plastic Plugged Sterile Sterilin Individually Paper Peel Wrapped 4 Packs of 50 ideal for tissue c ulture work 1 * 200 items
ALPCRC1030
:
Alpha Laboratories
Serological Pipette Graduated 5ml Plastic Plugged Sterile Sterilin Individually Paper Peel Wrapped 4 Packs of 50 ideal for tissue c ulture work 1 * 200 items
ALPCRC1030
:
Alpha Laboratories
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
Turbidity - WP, Certified Reference Material, Supelco®
: Merck
TRANSFER PIPETTE 5ML GRAD 0.25ML STERILE 1 * 3.000 items
: DELTALAB