Titan3 PTFE (Hydrophobic) Syringe Filter 0,45 um, 30 mm, yellow 1 * 100 items
Catalog # HYPE44525-NP
Supplier: Thermo Fisher Scientific
Titan3 PTFE (Hydrophobic) Syringe Filter 0,45 um, 30 mm, yellow 1 * 100 items
Catalog # HYPE44525-NP
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
Combination pH micro electrode, Orion™ PerpHecT™ ROSS™
Supplier: Thermo Orion
Accessories for diaphragm pumps
Supplier: BUCHI