ContactUsLinkComponent
Precision thermometer, enclosed scale, cap. prism. colourless, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, c. 300x9-10mm long, Richter-top finish with button, with Works Certificate with 1 test point 1 * 1 items
Catalog # AMARVWR48273
Supplier:  Amarell
Precision thermometer, enclosed scale, cap. prism. colourless, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, c. 300x9-10mm long, Richter-top finish with button, with Works Certificate with 1 test point 1 * 1 items
Catalog # AMARVWR48273
Supplier:  Amarell
Frequently Bought Together