ContactUsLinkComponent
Tube Vacutainer 2,7ml 13x75mm Trisodium Citrate 3,2% Unique Specimen Identification (USI) sterile 1 * 100 items
Catalog # BDAM363046
Supplier:  BD Medical
Tube Vacutainer 2,7ml 13x75mm Trisodium Citrate 3,2% Unique Specimen Identification (USI) sterile 1 * 100 items
Catalog # BDAM363046
Supplier:  BD Medical
Frequently Bought Together