ContactUsLinkComponent
Stainless steel plate stand with two support rods (height 700 mm), height adjustable drive unit holder and safety positioning bar. For more safety, the drive unit cannot be twisted sidewise or rotated. 1 * 1 items
Catalog # KINE11040052
Supplier:  KINEMATICA
Stainless steel plate stand with two support rods (height 700 mm), height adjustable drive unit holder and safety positioning bar. For more safety, the drive unit cannot be twisted sidewise or rotated. 1 * 1 items
Catalog # KINE11040052
Supplier:  KINEMATICA
Frequently Bought Together