ContactUsLinkComponent
MES monohydrate PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture 1 * 100 g
Catalog # SIALRES0113M-A701X
Supplier:  Merck
MES monohydrate PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture 1 * 100 g
Catalog # SIALRES0113M-A701X
Supplier:  Merck
Frequently Bought Together