2-Phenoxyethanol
Bottle, 25 g
Catalog # APOSOR4481-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
- CAS Number:122-99-6
- Formula:C₆H₅OCH₂CH₂OH
- MDL Number:MFCD00002857
- Molecular Weight:138.17 g/mol
- Boiling Pt:237…246 °C
- Melting Pt:41944 °C
- Density:1.107 g/cm³ (25 °C)
- Flash Pt:121 °C
- Storage Temperature:Ambient
- EINECS:204-589-7
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g



