Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
ɑ-Cyanocinnamic acid 95%

ɑ-Cyanocinnamic acid 95%

Bottle, 5 g
Catalog # APOSOR922724-5G
Supplier:   Apollo Scientific
ɑ-Cyanocinnamic acid 95%
Catalog # APOSOR922724-5G
Supplier:  Apollo Scientific
CAS Number:  1011-92-3
ɑ-Cyanocinnamic acid 95%

Bottle, 5 g

CAS Number:  1011-92-3
Price
1 770,00 Kč
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Cyano-3-phenylacrylic Acid, α-Cyanocinnamic acid, ['2-Cyano-3-phenylacrylic acid']
  • CAS Number:1011-92-3
  • Formula:C₁₀H₇NO₂
  • MDL Number:MFCD00016813
  • Molecular Weight:173.17 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications