ContactUsLinkComponent
Art.No. VWR47064 Special fabrication Hydrometer for propylene glycol, without thermometer, double scale, 0-60 in 10 vol. %, anti-freeze 0C to -51C, c. 255mm long 1 * 1 items
Catalog # AMARVWR47064
Supplier:  Amarell
Art.No. VWR47064 Special fabrication Hydrometer for propylene glycol, without thermometer, double scale, 0-60 in 10 vol. %, anti-freeze 0C to -51C, c. 255mm long 1 * 1 items
Catalog # AMARVWR47064
Supplier:  Amarell
Frequently Bought Together