VWR®, LED-Fluorescence Cubes
Blue LED fluorescence cube for TL534B FL, LED emission 460 nm,
Excitation filter 455-495 nm,
Dichroic mirror cut-off 500 nm,
Emission filter 510 LP nm
Excitation filter 455-495 nm,
Dichroic mirror cut-off 500 nm,
Emission filter 510 LP nm
Catalog # 630-3245
Supplier: VWR Collection
Jump to:
Product Details & Documents
For use with microscope series TL534B FL.
Documents
VWR® for Microscopy
Product Literature
Specifications
- Description:Blue LED fluorescence cube for TL534B FL
- Optical features:LED emission 460 nm,
Excitation filter 455-495 nm,
Dichroic mirror cut-off 500 nm,
Emission filter 510 LP nm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



