ContactUsLinkComponent
Flow Indicator WR1-025GMW Nominal diameter: DN 25 Connection: Female thread G 1 Housing: Brass nickel-plated Pressure resistance: PN 16 For media: Liquids Wiper: NBR Seals: NBR Media temperature: 0 ... 100 C Environmental temperature: 0 ... 70 C 1 * 1 items
Catalog # GRSG949823
Supplier:  Greisinger
Flow Indicator WR1-025GMW Nominal diameter: DN 25 Connection: Female thread G 1 Housing: Brass nickel-plated Pressure resistance: PN 16 For media: Liquids Wiper: NBR Seals: NBR Media temperature: 0 ... 100 C Environmental temperature: 0 ... 70 C 1 * 1 items
Catalog # GRSG949823
Supplier:  Greisinger
Frequently Bought Together