ContactUsLinkComponent
SONOREX TECHNIK RM 16.2 UH, ultrasonic bath, 40 kHz, 230 V~(± 10%) 50/60 Hz, ID 325x300x210/202 mm 20 l, welded tank 2 mm s/s AISI 316 Ti timer 1-15 min 1 * 1 items
Catalog # BAND8204
SONOREX TECHNIK RM 16.2 UH, ultrasonic bath, 40 kHz, 230 V~(± 10%) 50/60 Hz, ID 325x300x210/202 mm 20 l, welded tank 2 mm s/s AISI 316 Ti timer 1-15 min 1 * 1 items
Catalog # BAND8204
Frequently Bought Together