ContactUsLinkComponent
CHROMABOND QuEChERS Mix III Diamino clean-up mix consisting of: - 600 mg MgSO4 - 100 mg CHROMABOND Diamino individually weighed and filled into 15 mL centrifuge tubes (PP) with screw cap (PE) pack of 50 1 * 50 items
Catalog # MANA730980
Supplier:  MACHEREY-NAGEL
CHROMABOND QuEChERS Mix III Diamino clean-up mix consisting of: - 600 mg MgSO4 - 100 mg CHROMABOND Diamino individually weighed and filled into 15 mL centrifuge tubes (PP) with screw cap (PE) pack of 50 1 * 50 items
Catalog # MANA730980
Supplier:  MACHEREY-NAGEL
Frequently Bought Together