ContactUsLinkComponent
CHROMABOND columns XTR volume: 70 mL, content of sorbent: 17 g kieselguhr material: PP, with PE filter elements pack of 30 1 * 30 items
Catalog # MANA730510
Supplier:  MACHEREY-NAGEL
CHROMABOND columns XTR volume: 70 mL, content of sorbent: 17 g kieselguhr material: PP, with PE filter elements pack of 30 1 * 30 items
Catalog # MANA730510
Supplier:  MACHEREY-NAGEL
Frequently Bought Together