ContactUsLinkComponent
EC HPLC column EC 150/2 NUCLEOSIL 100-3 C18 length: 150 mm, ID: 2 mm pack of 1 1 * 1 items
Catalog # MANA720949.20
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 150/2 NUCLEOSIL 100-3 C18 length: 150 mm, ID: 2 mm pack of 1 1 * 1 items
Catalog # MANA720949.20
Supplier:  MACHEREY-NAGEL
Frequently Bought Together